Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEP
Entrez Gene ID
828706
Gene Name
RNA-binding KH domain-containing protein
Gene Aliases
AT4G26000, F20B18.110, F20B18_110, PEPPER
Chromosome Location
Chr.4: 13197069 - 13199841 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118733.4 NP_194330.1 5 - 6 1420 90
GenBank mRNA AY056288.1 - 5 - 6 1243 90
AY099659.1 - 5 - 6 1237 90
AY114064.1 - 5 - 6 1209 90

View Full Product Details