Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CK1
Entrez Gene ID
828716
Gene Name
casein kinase 1
Gene Aliases
AT4G26100, CKL1, casein kinase 1
Chromosome Location
Chr.4: 13227450 - 13230797 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118743.3 NP_194340.1 12 - 13 1276 60
GenBank mRNA AY050784.1 - 12 - 13 1276 60
AY091299.1 - 12 - 13 987 60
AY943842.1 - 12 - 13 987 60

View Full Product Details