Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BGAL12
Entrez Gene ID
828720
Gene Name
beta-galactosidase 12
Gene Aliases
AT4G26140, F20B18.250, F20B18_250, beta-galactosidase 12
Chromosome Location
Chr.4: 13242968 - 13248213 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341777.1 NP_001329560.1 9 - 10 1131 83
NM_001341778.1 NP_001329554.1 9 - 10 1073 83
NM_001341779.1 NP_001329557.1 9 - 10 1073 83
NM_001341780.1 NP_001329558.1 9 - 10 1074 83
NM_001341781.1 NP_001329559.1 9 - 10 1074 83
NM_001341782.1 NP_001329555.1 9 - 10 1073 83
NM_001341783.1 NP_001329556.1 9 - 10 1073 83
NM_118747.3 NP_194344.2 9 - 10 1131 83
NM_179222.2 NP_849553.1 9 - 10 1360 83
GenBank mRNA AJ270308.1 - 9 - 10 970 83
AY099612.1 - 9 - 10 1073 83
BT000267.1 - 9 - 10 970 83

View Full Product Details