Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFK3
Entrez Gene ID
828733
Gene Name
phosphofructokinase 3
Gene Aliases
AT4G26270, T25K17.80, T25K17_80, phosphofructokinase 3
Chromosome Location
Chr.4: 13300778 - 13304440 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118760.3 NP_567742.1 10 - 11 1472 67
GenBank mRNA AY049245.1 - 10 - 11 1380 67
AY090267.1 - 10 - 11 1062 67

View Full Product Details