Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UNE17
Entrez Gene ID
828739
Gene Name
Subtilisin-like serine endopeptidase family protein
Gene Aliases
AT4G26330, ATSBT3.18, T25K17.140, T25K17_140, UNFERTILIZED EMBRYO SAC 17
Chromosome Location
Chr.4: 13320408 - 13323673 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341795.1 NP_001328493.1 4 - 5 1042 78
NM_118766.2 NP_567744.1 4 - 5 1084 78

View Full Product Details