Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATRBP45C
Entrez Gene ID
828808
Gene Name
RNA-binding (RRM/RBD/RNP motifs) family protein
Gene Aliases
AT4G27000, F10M23.340, F10M23_340
Chromosome Location
Chr.4: 13554604 - 13557915 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118834.4 NP_567764.1 2 - 3 931 122
GenBank mRNA AF370339.1 - 2 - 3 870 122
AY054487.1 - 2 - 3 862 122
AY062999.1 - 2 - 3 779 122
AY093292.1 - 2 - 3 779 122

View Full Product Details