Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATRBP45C
Entrez Gene ID
828808
Gene Name
RNA-binding (RRM/RBD/RNP motifs) family protein
Gene Aliases
AT4G27000, F10M23.340, F10M23_340
Chromosome Location
Chr.4: 13554604 - 13557915 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118834.4 NP_567764.1 6 - 7 1400 65
GenBank mRNA AF370339.1 - 6 - 7 1339 65
AY054487.1 - 6 - 7 1331 65
AY062999.1 - 6 - 7 1248 65
AY093292.1 - 6 - 7 1248 65

View Full Product Details