Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WES1
Entrez Gene ID
828834
Gene Name
Auxin-responsive GH3 family protein
Gene Aliases
AT4G27260, GH3.5, M4I22.70, M4I22_70
Chromosome Location
Chr.4: 13653579 - 13656285 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118860.5 NP_194456.1 2 - 3 1382 94
GenBank mRNA AY070038.1 - 2 - 3 1343 94
AY096516.1 - 2 - 3 1257 94
DQ512891.1 - 2 - 3 1257 94

View Full Product Details