Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTG2
Entrez Gene ID
828874
Gene Name
GPCR-type G protein 2
Gene Aliases
AT4G27630, GPCR-type G protein 2, T29A15.120, T29A15_120
Chromosome Location
Chr.4: 13791642 - 13797305 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203925.2 NP_001190854.1 5 - 6 796 82
NM_001341872.1 NP_001320077.1 4 - 5 1182 82
NM_001341873.1 NP_001329564.1 5 - 6 787 82
NM_001341874.1 NP_001329563.1 4 - 5 1182 82
NM_001341875.1 NP_001329565.1 5 - 6 787 82
NM_118899.4 NP_194493.2 5 - 6 945 82
NM_179224.2 NP_849555.1 2 - 3 637 82
GenBank mRNA HM776217.1 - 5 - 6 673 82

View Full Product Details