Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLE14
Entrez Gene ID
28717391
Gene Name
CLAVATA3/ESR-RELATED 14
Gene Aliases
AT1G63245, CLAVATA3/ESR-RELATED 14
Chromosome Location
Chr.1: 23460198 - 23460884 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334074.1 NP_001319300.1 1 - 1 301 68

View Full Product Details