Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP71A21
- Entrez Gene ID
-
823990
- Gene Name
- cytochrome P450, family 71, subfamily A, polypeptide 21
- Gene Aliases
-
''cytochrome P450, AT3G48320, cytochrome P450, family 71, polypeptide 21, polypeptide 21'', subfamily A
- Chromosome Location
-
Chr.3: 17891134 - 17892847 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_148858.2 |
NP_680111.1 |
1 - 2 |
959 |
88 |
View Full Product Details