Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl16
Entrez Gene ID
820402
Gene Name
serine carboxypeptidase-like 16
Gene Aliases
AT3G12220, serine carboxypeptidase-like 16
Chromosome Location
Chr.3: 3896207 - 3899019 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337982.1 NP_001327319.1 9 - 10 1119 128
NM_001337983.1 NP_001327318.1 8 - 9 890 128
NM_112058.4 NP_566414.3 10 - 11 1046 128

View Full Product Details