The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001337982.1 | NP_001327319.1 | 6 - 7 | 904 | 103 |
NM_112058.4 | NP_566414.3 | 7 - 8 | 831 | 103 |