Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSF2
Entrez Gene ID
820432
Gene Name
PSF2
Gene Aliases
AT3G12530
Chromosome Location
Chr.3: 3972180 - 3974440 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035603.1 NP_001030680.1 3 - 4 212 82
NM_001337995.1 NP_001319531.1 5 - 6 415 82
NM_001337996.1 NP_001326331.1 4 - 5 438 82
NM_112088.3 NP_187859.2 4 - 5 537 82
GenBank mRNA BT010931.1 - 4 - 5 296 82
BT020600.1 - 3 - 4 231 82

View Full Product Details