Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPGR2
Entrez Gene ID
828978
Gene Name
no pollen germination related 2
Gene Aliases
AT4G28600, T5F17.50, T5F17_50, no pollen germination related 2
Chromosome Location
Chr.4: 14129180 - 14132869 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341925.1 NP_001329155.1 5 - 6 2537 88
NM_001341926.1 NP_001329156.1 5 - 6 2348 88
NM_119002.6 NP_194589.2 5 - 6 2333 88
GenBank mRNA BT015369.1 - 5 - 6 2033 88
BT020340.1 - 4 - 5 1896 88

View Full Product Details