Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ckl3
Entrez Gene ID
829009
Gene Name
casein kinase I-like 3
Gene Aliases
AT4G28880, F25O24.3, casein kinase I-like 3
Chromosome Location
Chr.4: 14251135 - 14254946 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341951.1 NP_001329056.1 13 - 14 1087 65
NM_001341952.1 NP_001329055.1 12 - 13 1190 65
NM_001341953.1 NP_001329054.1 13 - 14 1083 65
NM_119032.5 NP_194617.2 12 - 13 1190 65
GenBank mRNA AK118601.1 - 12 - 13 1106 65
AY058842.1 - 12 - 13 1039 65
AY079031.1 - 12 - 13 974 65
AY943852.1 - 12 - 13 974 65

View Full Product Details