Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATMND1
Entrez Gene ID
829038
Gene Name
Mnd1 family protein
Gene Aliases
AT4G29170, F19B15.200, F19B15_200
Chromosome Location
Chr.4: 14382819 - 14385465 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085000.2 NP_001078469.1 6 - 7 649 81
NM_001341959.1 NP_001329788.1 6 - 7 571 81
NM_119061.4 NP_194646.2 6 - 7 554 81
GenBank mRNA AK117713.1 - 6 - 7 503 81
AM162278.1 - 5 - 6 405 81
BT005435.1 - 5 - 6 405 81
DQ248000.1 - 6 - 7 526 81

View Full Product Details