Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFK1
Entrez Gene ID
829043
Gene Name
phosphofructokinase 1
Gene Aliases
AT4G29220, F17A13.40, F17A13_40, phosphofructokinase 1
Chromosome Location
Chr.4: 14403347 - 14406592 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119066.3 NP_194651.1 10 - 11 1446 105
GenBank mRNA BT015793.1 - 9 - 10 1074 105

View Full Product Details