Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRMT11
Entrez Gene ID
829072
Gene Name
arginine methyltransferase 11
Gene Aliases
ARABIDOPSIS THALIANA ARGININE METHYLTRANSFERASE 11, ARABIDOPSIS THALIANA PROTEIN ARGININE METHYLTRANSFERASE 1B, ARABIDOPSIS THALIANA PROTEIN ARGININE METHYLTRANSFERASE 1B, AT4G29510, ATPRMT11, ATPRMT1B, PRMT1B, PROTEIN ARGININE METHYLTRANSFERASE 1B, T16L4.20, T16L4_20, arginine methyltransferase 11
Chromosome Location
Chr.4: 14491622 - 14494001 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119096.3 NP_194680.1 7 - 8 929 79
GenBank mRNA AY087817.1 - 7 - 8 926 79
AY150407.1 - 7 - 8 812 79

View Full Product Details