Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CFIM-25
Entrez Gene ID
829104
Gene Name
homolog of CFIM-25
Gene Aliases
ARABIDOPSIS THALIANA HOMOLOG OF CFIM-25, AT4G29820, ATCFIM-25, F27B13.60, F27B13_60, homolog of CFIM-25
Chromosome Location
Chr.4: 14595680 - 14597516 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119128.3 NP_567835.1 6 - 7 799 72
GenBank mRNA AY048229.1 - 6 - 7 762 72
AY085984.1 - 6 - 7 764 72
AY091704.1 - 6 - 7 600 72

View Full Product Details