Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PRHA
- Entrez Gene ID
-
829117
- Gene Name
- pathogenesis related homeodomain protein A
- Gene Aliases
-
AT4G29940, F27B13.180, F27B13_180, HOMEODOMAIN PROTEIN, pathogenesis related homeodomain protein A, pathogenesis related homeodomain protein A
- Chromosome Location
-
Chr.4: 14648138 - 14653489 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119140.2 |
NP_194723.1 |
11 - 12 |
2191 |
71 |
GenBank mRNA |
L21991.1 |
- |
11 - 12 |
2106 |
71 |
View Full Product Details