The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001341993.1 | NP_001320088.1 | 8 - 9 | 1615 | 163 |
NM_119157.4 | NP_194740.1 | 7 - 8 | 1470 | 163 |