Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMA3
Entrez Gene ID
829135
Gene Name
heavy metal atpase 3
Gene Aliases
A. THALIANA HEAVY METAL ATPASE 3, AT4G30120, ATHMA3, F6G3.150, F6G3_150, heavy metal atpase 3
Chromosome Location
Chr.4: 14730215 - 14733628 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119158.4 NP_194741.2 1 - 2 330 95
GenBank mRNA DQ446885.1 - 1 - 2 204 95

View Full Product Details