Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HA2
Entrez Gene ID
829142
Gene Name
H[+]-ATPase 2
Gene Aliases
AHA2, AT4G30190, F9N11.40, F9N11_40, H(+)-ATPase 2, P-TYPE H(+)-ATPASE ISOFORM 2, PLASMA MEMBRANE PROTON ATPASE 2, PMA2
Chromosome Location
Chr.4: 14770395 - 14776247 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203941.2 NP_001190870.1 2 - 3 307 99
NM_119165.4 NP_194748.1 2 - 3 496 99
GenBank mRNA AY035075.1 - 2 - 3 297 99
BT000781.1 - 2 - 3 297 99
BT001969.1 - 2 - 3 169 99

View Full Product Details