Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RUXF
Entrez Gene ID
829145
Gene Name
small nuclear ribonucleoprotein F
Gene Aliases
AT4G30220, F9N11.70, F9N11_70, small nuclear ribonucleoprotein F
Chromosome Location
Chr.4: 14802931 - 14804461 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203943.1 NP_001190872.1 4 - 5 322 61
NM_119168.4 NP_194751.1 4 - 5 401 61
GenBank mRNA AK227757.1 - 4 - 5 232 61
AY087616.1 - 4 - 5 263 61
BT006382.1 - 3 - 4 199 61

View Full Product Details