Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XTH24
Entrez Gene ID
829150
Gene Name
xyloglucan endotransglucosylase/hydrolase 24
Gene Aliases
AT4G30270, F9N11.120, F9N11_120, MERI-5, MERI5B, MERISTEM 5, SEN4, SENESCENCE 4, meristem-5, xyloglucan endotransglucosylase/hydrolase 24
Chromosome Location
Chr.4: 14819192 - 14820595 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119173.4 NP_194756.1 1 - 2 409 153
GenBank mRNA AK229363.1 - 1 - 2 321 153
AK230296.1 - - 1386 153
AY035156.1 - 1 - 2 321 153
AY063027.1 - 1 - 2 262 153
D63508.1 - 1 - 2 293 153

View Full Product Details