Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCE3
- Entrez Gene ID
-
829153
- Gene Name
- non-intrinsic ABC protein 15
- Gene Aliases
-
ARABIDOPSIS THALIANA NON-INTRINSIC ABC PROTEIN 15, AT4G30300, ATNAP15, ATP-binding cassette E3, F17I23.360, F17I23_360, NAP15, non-intrinsic ABC protein 15
- Chromosome Location
-
Chr.4: 14831005 - 14831550 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119176.1 |
NP_194759.1 |
1 - 1 |
498 |
90 |
GenBank mRNA |
DQ056666.1 |
- |
1 - 1 |
498 |
90 |
View Full Product Details