Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNGC9
Entrez Gene ID
829179
Gene Name
cyclic nucleotide gated channel 9
Gene Aliases
AT4G30560, ATCNGC9, F17I23.100, F17I23_100, cyclic nucleotide gated channel 9
Chromosome Location
Chr.4: 14926834 - 14930355 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203944.2 NP_001190873.1 4 - 5 1140 80
NM_001342012.1 NP_001329898.1 3 - 4 1269 80
NM_001342013.1 NP_001329900.1 4 - 5 1257 80
NM_001342014.1 NP_001329899.1 2 - 3 856 80
NM_119202.4 NP_194785.1 4 - 5 1121 80

View Full Product Details