Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAUT12
Entrez Gene ID
835558
Gene Name
galacturonosyltransferase 12
Gene Aliases
AT5G54690, IRREGULAR XYLEM 8, IRX8, K5F14.3, K5F14_3, LGT6, galacturonosyltransferase 12
Chromosome Location
Chr.5: 22219224 - 22221968 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124850.4 NP_200280.1 3 - 4 1222 115
GenBank mRNA AK229580.1 - 3 - 4 1094 115
BT023729.1 - 3 - 4 1089 115
KJ138638.1 - 3 - 4 1023 115

View Full Product Details