Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBP45A
Entrez Gene ID
835581
Gene Name
RNA-binding protein 45A
Gene Aliases
AT5G54900, ATRBP45A, RNA-binding protein 45A
Chromosome Location
Chr.5: 22295369 - 22298395 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124872.3 NP_568815.1 4 - 5 958 76
GenBank mRNA AF324983.2 - 4 - 5 944 76
BT029545.1 - 4 - 5 915 76

View Full Product Details