Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EDM2
Entrez Gene ID
835632
Gene Name
ENHANCED DOWNY MILDEW 2
Gene Aliases
AT5G55390, ENHANCED DOWNY MILDEW 2, MTE17.10, MTE17_10
Chromosome Location
Chr.5: 22447937 - 22454805 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203616.2 NP_001190545.1 9 - 10 1365 78
NM_001345123.1 NP_001332152.1 8 - 9 1633 78
NM_124921.5 NP_200350.2 9 - 10 1420 78
GenBank mRNA KF112069.1 - 8 - 9 1242 78

View Full Product Details