Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GEX1
Entrez Gene ID
835642
Gene Name
gamete expressed protein 1
Gene Aliases
AT5G55490, ATGEX1, GAMETE EXPRESSED PROTEIN 1, MTE17.20, MTE17_20, gamete expressed protein 1
Chromosome Location
Chr.5: 22478712 - 22481937 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345127.1 NP_001318802.1 5 - 6 1461 136
NM_001345130.1 NP_001330004.1 4 - 5 1570 136
NM_001345133.1 NP_001330005.1 5 - 6 1483 136
NM_124931.2 NP_200360.2 4 - 5 1457 136
GenBank mRNA AK175610.1 - 4 - 5 1457 136

View Full Product Details