Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRT1
Entrez Gene ID
835670
Gene Name
sirtuin 1
Gene Aliases
AT5G55760, sirtuin 1
Chromosome Location
Chr.5: 22567222 - 22571008 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124958.3 NP_200387.1 13 - 14 1171 86
GenBank mRNA AF283757.1 - 13 - 14 1131 86

View Full Product Details