Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NMNAT
- Entrez Gene ID
-
835675
- Gene Name
- nicotinate/nicotinamide mononucleotide adenyltransferase
- Gene Aliases
-
AT5G55810, AtNMNAT, MDF20.25, MDF20_25, nicotinate/nicotinamide mononucleotide adenyltransferase
- Chromosome Location
-
Chr.5: 22583972 - 22586405 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124963.5 |
NP_200392.3 |
4 - 5 |
463 |
115 |
View Full Product Details