Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERD3
Entrez Gene ID
827650
Gene Name
S-adenosyl-L-methionine-dependent methyltransferases superfamily protein
Gene Aliases
AT4G19120, T18B16.90, T18B16_90, early-responsive to dehydration 3
Chromosome Location
Chr.4: 10460241 - 10464306 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341325.1 NP_001329718.1 3 - 4 697 101
NM_118031.4 NP_567575.1 3 - 4 765 101
NM_179077.2 NP_849408.1 2 - 3 1369 101
GenBank mRNA AB039927.1 - 3 - 4 621 101
AK317548.1 - 2 - 3 471 101
AK317670.1 - 3 - 4 633 101

View Full Product Details