The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197982.1 | NP_001184911.1 | 12 - 13 | 2213 | 87 |
NM_001331528.1 | NP_001322680.1 | 11 - 12 | 2203 | 87 | |
NM_001331529.1 | NP_001322678.1 | 10 - 11 | 2070 | 87 | |
NM_001331531.1 | NP_001322673.1 | 10 - 11 | 3290 | 87 | |
NM_001331532.1 | NP_001322674.1 | 11 - 12 | 3199 | 87 | |
NM_001331533.1 | NP_001322675.1 | 11 - 12 | 3172 | 87 | |
NM_001331534.1 | NP_001322676.1 | 9 - 10 | 2877 | 87 | |
NM_001331535.1 | NP_001322677.1 | 10 - 11 | 2786 | 87 | |
NM_100402.3 | NP_172015.1 | 10 - 11 | 2759 | 87 | |
NM_179265.2 | NP_849596.1 | 11 - 12 | 2222 | 87 | |
GenBank mRNA | AK316776.1 | - | 11 - 12 | 2127 | 87 |
AY037177.1 | - | 10 - 11 | 1916 | 87 | |
AY091690.1 | - | 9 - 10 | 1812 | 87 |