Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDG2
Entrez Gene ID
839256
Gene Name
homeodomain GLABROUS 2
Gene Aliases
AT1G05230, YUP8H12.16, YUP8H12_16, homeodomain GLABROUS 2
Chromosome Location
Chr.1: 1512896 - 1518900 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123759.1 NP_001117231.1 2 - 3 343 63
NM_001197982.1 NP_001184911.1 4 - 5 638 63
NM_001331528.1 NP_001322680.1 3 - 4 628 63
NM_001331529.1 NP_001322678.1 2 - 3 495 63
NM_001331530.1 NP_001322679.1 3 - 4 541 63
NM_001331531.1 NP_001322673.1 2 - 3 1715 63
NM_001331532.1 NP_001322674.1 3 - 4 1624 63
NM_001331533.1 NP_001322675.1 3 - 4 1597 63
NM_001331534.1 NP_001322676.1 1 - 2 1302 63
NM_001331535.1 NP_001322677.1 2 - 3 1211 63
NM_100402.3 NP_172015.1 2 - 3 1184 63
NM_179265.2 NP_849596.1 3 - 4 647 63
GenBank mRNA AK316776.1 - 3 - 4 552 63
AY037177.1 - 2 - 3 341 63
AY091690.1 - 1 - 2 237 63

View Full Product Details