Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDG2
Entrez Gene ID
839256
Gene Name
homeodomain GLABROUS 2
Gene Aliases
AT1G05230, YUP8H12.16, YUP8H12_16, homeodomain GLABROUS 2
Chromosome Location
Chr.1: 1512896 - 1518900 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123759.1 NP_001117231.1 7 - 8 1379 84
NM_001197982.1 NP_001184911.1 9 - 10 1674 84
NM_001331528.1 NP_001322680.1 8 - 9 1664 84
NM_001331529.1 NP_001322678.1 7 - 8 1531 84
NM_001331530.1 NP_001322679.1 8 - 9 1577 84
NM_001331531.1 NP_001322673.1 7 - 8 2751 84
NM_001331532.1 NP_001322674.1 8 - 9 2660 84
NM_001331533.1 NP_001322675.1 8 - 9 2633 84
NM_001331534.1 NP_001322676.1 6 - 7 2338 84
NM_001331535.1 NP_001322677.1 7 - 8 2247 84
NM_100402.3 NP_172015.1 7 - 8 2220 84
NM_179265.2 NP_849596.1 8 - 9 1683 84
GenBank mRNA AK316776.1 - 8 - 9 1588 84
AY037177.1 - 7 - 8 1377 84
AY091690.1 - 6 - 7 1273 84

View Full Product Details