Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLX2-4
Entrez Gene ID
837122
Gene Name
glyoxalase 2-4
Gene Aliases
AT1G06130, T21E18.18, T21E18_18, glyoxalase 2-4
Chromosome Location
Chr.1: 1857757 - 1860938 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100494.5 NP_563760.1 6 - 7 1043 80
NM_179268.2 NP_849599.1 6 - 7 800 80
GenBank mRNA AK227097.1 - 6 - 7 757 80
AY085754.1 - 6 - 7 817 80
AY099584.1 - 6 - 7 750 80
AY128833.1 - 6 - 7 742 80

View Full Product Details