The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001331755.1 | NP_001322007.1 | 6 - 7 | 2979 | 141 |
NM_100680.4 | NP_563806.1 | 6 - 7 | 2976 | 141 | |
NM_179277.1 | NP_849608.1 | 5 - 6 | 2708 | 141 |