Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAMMA-ADAPTIN 1
Entrez Gene ID
839001
Gene Name
gamma-adaptin 1
Gene Aliases
AT1G23900, Gamma-ADR, Gamma-adaptin, gamma-adaptin 1
Chromosome Location
Chr.1: 8441073 - 8447681 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332624.1 NP_001322140.1 16 - 17 2995 116
NM_102238.2 NP_173802.1 17 - 18 2575 116
NM_179370.4 NP_849701.1 16 - 17 2732 116
GenBank mRNA BT004307.1 - 16 - 17 2648 116
BT005557.1 - 16 - 17 2432 116

View Full Product Details