The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_112103.3 | NP_187874.2 | 10 - 11 | 1418 | 65 |
GenBank mRNA | AY024357.1 | - | 10 - 11 | 1418 | 65 |