The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_112125.3 | NP_566437.1 | 1 - 2 | 370 | 79 |
GenBank mRNA | AY085594.1 | - | 1 - 2 | 372 | 79 |
AY885244.1 | - | 1 - 2 | 346 | 79 |