Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GIP1
Entrez Gene ID
820513
Gene Name
GBF-interacting protein 1
Gene Aliases
AT3G13222, GBF-interacting protein 1
Chromosome Location
Chr.3: 4250958 - 4254278 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112163.6 NP_187929.4 4 - 5 491 130
GenBank mRNA AY063883.1 - 4 - 5 450 130
AY142588.1 - 4 - 5 419 130

View Full Product Details