Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BGAL1
Entrez Gene ID
820584
Gene Name
beta galactosidase 1
Gene Aliases
AT3G13750, beta galactosidase 1, beta-galactosidase 1
Chromosome Location
Chr.3: 4510965 - 4516478 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112225.5 NP_187988.1 14 - 15 1896 102
GenBank mRNA AJ270297.1 - 14 - 15 1874 102

View Full Product Details