Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAP1;4
Entrez Gene ID
820589
Gene Name
nucleosome assembly protein1;4
Gene Aliases
AT3G13782, NFA04, NFA4, NUCLEOSOME/CHROMATIN ASSEMBLY FACTOR GROUP A 04, NUCLEOSOME/CHROMATIN ASSEMBLY FACTOR GROUP A 4, nucleosome assembly protein1;4
Chromosome Location
Chr.3: 4526220 - 4528646 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338075.1 NP_001319544.1 7 - 8 812 116
NM_112230.3 NP_187993.2 6 - 7 693 116

View Full Product Details