Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CWINV5
Entrez Gene ID
820590
Gene Name
cell wall invertase 5
Gene Aliases
AT3G13784, AtcwINV5, cell wall invertase 5
Chromosome Location
Chr.3: 4528362 - 4530713 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112231.2 NP_187994.1 1 - 2 194 93

View Full Product Details