Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CWINV5
Entrez Gene ID
820590
Gene Name
cell wall invertase 5
Gene Aliases
AT3G13784, AtcwINV5, cell wall invertase 5
Chromosome Location
Chr.3: 4528362 - 4530713 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338076.1 NP_001326013.1 4 - 5 1581 135
NM_001338077.1 NP_001326012.1 3 - 4 1395 135
NM_001338078.1 NP_001326014.1 3 - 4 1537 135
NM_112231.2 NP_187994.1 3 - 4 1556 135

View Full Product Details