Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSP60-3A
Entrez Gene ID
820599
Gene Name
heat shock protein 60-3A
Gene Aliases
AT3G13860, heat shock protein 60-3A
Chromosome Location
Chr.3: 4561517 - 4565335 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112240.4 NP_566466.1 7 - 8 929 139
GenBank mRNA AY094453.1 - 7 - 8 843 139
BT015923.1 - 7 - 8 727 139

View Full Product Details