Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHD3
Entrez Gene ID
820600
Gene Name
Root hair defective 3 GTP-binding protein (RHD3)
Gene Aliases
AT3G13870, GOLGI MUTANT 8, GOM8, ROOT HAIR DEFECTIVE 3
Chromosome Location
Chr.3: 4565370 - 4571355 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112241.4 NP_188003.1 11 - 12 1088 75
NM_202579.1 NP_974308.1 11 - 12 997 75
GenBank mRNA AY092988.1 - 11 - 12 932 75
AY128801.1 - 10 - 11 842 75

View Full Product Details